3-Quinolineboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinoline-3-boronic acid, Quinolin-3-ylboronic acid

L20088-06 L20088-MD L20088-03
AAAL20088-06EA 154 USD
AAAL20088-06 AAAL20088-MD AAAL20088-03
3-Quinolineboronic acid 95%
3-Quinolineboronic acid

Purity: 95% CAS No.: 191162-39-7 Molecular Formula: C9H8BNO2 Formula Weight: 172.98 Melting Point: 300° Beilstein Reference: 8764547

Formula: C₉H₈BNO₂
MW: 172.98 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD02183527
CAS Number: 191162-39-7
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR