2-Fluoro-4-methoxyphenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Fluoro-4-methoxybenzeneboronic acid
L19960-06
L19960-03
AAAL19960-06EA
225
USD
AAAL19960-06
AAAL19960-03
2-Fluoro-4-methoxyphenylboronic acid 98%
2-Fluoro-4-methoxyphenylboronic acid
Purity: 98%
CAS No.: 162101-31-7
Molecular Formula: C7H8BFO3
Formula Weight: 169.95
Beilstein Reference: 9192698
MDL No.: MFCD04039323
Formula:
C₇H₈BFO₃ MW: 169.95 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04039323 CAS Number: 162101-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...