8-Quinolineboronic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinolin-8-ylboronic acid

L19640-03 L19640-MD
AAAL19640-03EA 137 USD
AAAL19640-03 AAAL19640-MD
8-Quinolineboronic acid 99%
8-Quinolineboronic acid

Purity: 99% CAS No.: 86-58-8 Molecular Formula: C9H8BNO2 Formula Weight: 172.98 Melting Point: 300° Beilstein Reference: 147522 Merck Reference: 14,8069 MDL No.: MFCD01114698

Formula: C₉H₈BNO₂
MW: 172.98 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01114698
CAS Number: 86-58-8
UN: 2811
ADR: "6.1",III
Merck Index: 14,8069

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR