5-Bromoindoline 98+%

Supplier: Thermo Scientific Chemicals
Warning

L16992-06 L16992-03
AAAL16992-06EA 137 USD
AAAL16992-06 AAAL16992-03
5-Bromoindoline 98+%
5-Bromoindoline

Purity: 98+% CAS No.: 22190-33-6 Molecular Formula: C8H8BrN Formula Weight: 198.06 Melting Point: 36-40° Boiling Point: 114-115°/5mm MDL No.: MFCD00027410

Formula: C₈H₈BrN
MW: 198.06 g/mol
Boiling Pt: 114…115 °C/5 mm
Melting Pt: 36…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00027410
CAS Number: 22190-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR