Tantalum(V) ethanolate 99.95% (metals basis), Optical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tantalum(V) ethoxide

L16033-03 L16033-06
AAAL16033-03EA 34 USD
AAAL16033-03 AAAL16033-06
Tantalum(V) ethanolate 99.95% (metals basis), Optical Grade
Tantalum(V) ethanolate

Purity: 99.95% CAS No.: 6074-84-6 Molecular Formula: C10H25O5Ta Formula Weight: 406.25 Flash Point: 30°(86°F) Refractive Index: 1.4880 Melting Point: 18-19° Boiling Point: 145°/0.1mm Beilstein Reference: 3678999 UN No.: 2924 MDL No.: MFCD00049785

Formula: C₁₀H₂₅O₅Ta
MW: 406.25 g/mol
Boiling Pt: 145 °C/0.1 mm
Melting Pt: 18…19 °C
Density: 1.566
Flash Pt: 30 °C(86 °F)
Storage Temperature: Ambient
MDL Number: MFCD00049785
CAS Number: 6074-84-6
EINECS: 228-010-2
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR