Ethanolamine hydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Aminoethanol hydrochloride, 2-Hydroxyethylammonium Chloride, Monoethanolamine Hydrochloride, 2-Aminoethan-1-ol hydrochloride

L14322-22 L14322-36
AAAL14322-22EA 49.2 USD
AAAL14322-22 AAAL14322-36
Ethanolamine hydrochloride 98+%
Ethanolamine hydrochloride

Purity: 98+% CAS No.: 2002-24-6 Molecular Formula: HOCH2CH2NH2.HCl Formula Weight: 97.55 Melting Point: 83-86° Beilstein Reference: 3542892 Merck Reference: 14,3727 MDL No.: MFCD00012892

Formula: C₂H₈ClNO
MW: 97.54 g/mol
Melting Pt: 83…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00012892
CAS Number: 2002-24-6
EINECS: 217-900-6
Merck Index: 14,3727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR