p-Phenylenediacetonitrile 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2'-(1,4-Phenylene)diacetonitrile, 1,4-Phenylenediacetonitrile

L13464-14 L13464-06
AAAL13464-14EA 168 USD
AAAL13464-14 AAAL13464-06
p-Phenylenediacetonitrile 97%
p-Phenylenediacetonitrile

Purity: 97% CAS No.: 622-75-3 Molecular Formula: C10H8N2 Formula Weight: 156.19 Melting Point: 94-98° Beilstein Reference: 1866157 UN No.: 3439 MDL No.: MFCD00001923

Formula: C₁₀H₈N₂
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00001923
CAS Number: 622-75-3
EINECS: 210-751-8
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR