Decylphosphonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: n-Decylphosphonic acid, 1-Decylphosphonic acid

L12019-03 L12019-06
AAAL12019-03EA 55.3 USD
AAAL12019-03 AAAL12019-06
Decylphosphonic acid 98%
Decylphosphonic acid

Purity: 98% CAS No.: 6874-60-8 Molecular Formula: CH3(CH2)9PO3H2 Formula Weight: 222.27 Melting Point: 100-101° Beilstein Reference: 1771572 UN No.: 3261 MDL No.: MFCD00015830

Formula: C₁₀H₂₃O₃P
MW: 222.26 g/mol
Melting Pt: 100…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00015830
CAS Number: 6874-60-8
EINECS: 229-975-2
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR