3-Furoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11629-14
L11629-06
AAAL11629-14EA
309
USD
AAAL11629-14
AAAL11629-06
3-Furoic acid 99%
3-Furoic acid
Purity: 99%
CAS No.: 488-93-7
Molecular Formula: C5H4O3
Formula Weight: 112.08
Melting Point: 120-123°
Beilstein Reference: 108638
Formula:
C₅H₄O₃ Melting Pt: 120…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005350 CAS Number: 488-93-7 EINECS: 207-689-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...