Methyltriphenylsilane 97%
Supplier: Thermo Scientific Chemicals
L10995-03
L10995-06
L10995-14
AAAL10995-03EA
32.2
USD
AAAL10995-03
AAAL10995-06
AAAL10995-14
Methyltriphenylsilane 97%
Methyltriphenylsilane
Purity: 97%
CAS No.: 791-29-7
Molecular Formula: C19H18Si
Formula Weight: 274.44
Melting Point: 66-68°
Beilstein Reference: 2940360
Formula:
C₁₉H₁₈Si MW: 274.44 g/mol Melting Pt: 66…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041325 CAS Number: 791-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...