(Aminomethyl)phosphonic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (Aminomethyl)phosphonic Acid, AMPA

L09833-MD L09833-03
AAAL09833-MDEA 56.6 USD
AAAL09833-MD AAAL09833-03
(Aminomethyl)phosphonic acid 99%
(Aminomethyl)phosphonic acid

Purity: 99% CAS No.: 1066-51-9 Molecular Formula: CH6NO3P Formula Weight: 111.04 Melting Point: ca 290° dec. Beilstein Reference: 1701219

Formula: CH₆NO₃P
MW: 111.04 g/mol
Melting Pt: ca 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008105
CAS Number: 1066-51-9
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR