β-Phenylpropiophenone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: beta-Phenylpropiophenone, 1,3-Diphenyl-1-propanone

L09173-09 L09173-18
AAAL09173-09EA 115 USD
AAAL09173-09 AAAL09173-18
β-Phenylpropiophenone 98%
β-Phenylpropiophenone

Purity: 98% CAS No.: 1083-30-3 Molecular Formula: C15H14O Formula Weight: 210.28 Melting Point: 68-73° Beilstein Reference: 1910661

Formula: C₁₅H₁₄O
MW: 210.28 g/mol
Melting Pt: 68…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00039563
CAS Number: 1083-30-3
EINECS: 621-467-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR