α,α-Dimethylpropiophenone 98%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
2,2,2-Trimethylacetophenone
L08729-03
L08729-14
L08729-06
AAAL08729-03EA
54.1
USD
AAAL08729-03
AAAL08729-14
AAAL08729-06
α,α-Dimethylpropiophenone 98%
α,α-Dimethylpropiophenone
Purity: 98%
CAS No.: 938-16-9
Molecular Formula: C11H14O
Formula Weight: 162.23
Flash Point: 87°(188°F)
Refractive Index: 1.5084
Boiling Point: 219-222°
Beilstein Reference: 1906460
MDL No.: MFCD00008844
Formula:
C₁₁H₁₄O MW: 162.23 g/mol Boiling Pt: 219…222 °C Density: 0,970 Flash Pt: 87 °C(188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008844 CAS Number: 938-16-9 EINECS: 213-338-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...