Diphenyl phosphoramidate 97%
Supplier: Thermo Scientific Chemicals
L07155-06
L07155-14
AAAL07155-06EA
66.6
USD
AAAL07155-06
AAAL07155-14
Diphenyl phosphoramidate 97%
Diphenyl phosphoramidate
Purity: 97%
CAS No.: 2015-56-7
Molecular Formula: C12H12NO3P
Formula Weight: 249.21
Melting Point: 147-151°
Beilstein Reference: 2054166
Formula:
C₁₂H₁₂NO₃P |
MDL Number:
MFCD00014078 CAS Number: 2015-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...