Methyl-5-amino-2-furoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 5-aminofuran-2-carboxylate

L05958-03 L05958-06
AAAL05958-03EA 102 USD
AAAL05958-03 AAAL05958-06
Methyl-5-amino-2-furoate 98%
Methyl-5-amino-2-furoate

Purity: 98% CAS No.: 22600-30-2 Molecular Formula: C6H7NO3 Formula Weight: 141.13 Melting Point: 131-135° Beilstein Reference: 1365799

Formula: C₆H₇NO₃
MW: 141.13 g/mol
Melting Pt: 131…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00051661
CAS Number: 22600-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR