Dichloro(phenyl)silane 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L04371-14
L04371-06
AAAL04371-14EA
227.54
USD
AAAL04371-14
AAAL04371-06
Dichloro(phenyl)silane 96%
Dichloro(phenyl)silane
Purity: 96%
CAS No.: 1631-84-1
Molecular Formula: C6H6Cl2Si
Formula Weight: 177.11
Flash Point: 48°(118°F)
Refractive Index: 1.5230
Boiling Point: 181°
Beilstein Reference: 2433865
UN No.: 2986
MDL No.: MFCD00039296
Formula:
C₆H₆Cl₂Si MW: 177.1 g/mol Boiling Pt: 181 °C Density: 1.212 Flash Pt: 48 °C(118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039296 CAS Number: 1631-84-1 EINECS: 216-635-3 UN: 2986 ADR: "8",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...