Ethyl-1,3-dithiane-2-carboxylate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Fieser: 4,230
Purity: 98+%
CAS No.: 20462-00-4
Molecular Formula: C7H12O2S2
Formula Weight: 192.30
Flash Point: 54°(129°F)
Refractive Index: 1.5400
Melting Point: 19-21°
Boiling Point: 75-77°/0.2mm
Beilstein Reference: 1424352
UN No.: 1993
MDL No.: MFCD00006657
Formula:
C₇H₁₂O₂S₂ Boiling Pt: 75…77 °C/0.2 mm Melting Pt: 19…21 °C Density: 1.220 Flash Pt: 54 °C(129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006657 CAS Number: 20462-00-4 EINECS: 243-838-4 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...