4-Iodobiphenyl 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Iodo-1,1'-biphenyl

H27453-03 H27453-06 H27453-14
AAH27453-03EA 45.3 USD
AAH27453-03 AAH27453-06 AAH27453-14
4-Iodobiphenyl 97%
4-Iodobiphenyl

Purity: 97% CAS No.: 1591-31-7 Molecular Formula: C12H9I Formula Weight: 280.11 Melting Point: 110-114° UN No.: 3077 MDL No.: MFCD00019028

Formula: C₁₂H₉I
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 110…114 °C
Density: 1.584 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00019028
CAS Number: 1591-31-7
EINECS: 216-469-1
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR