3-(Methoxycarbonyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-boronobenzoate, 3-(Methoxycarbonyl)benzeneboronic acid

H27444-06 H27444-03
AAH27444-06EA 93.4 USD
AAH27444-06 AAH27444-03
3-(Methoxycarbonyl)phenylboronic acid 97%
3-(Methoxycarbonyl)phenylboronic acid

Purity: 97% CAS No.: 99769-19-4 Molecular Formula: C8H9BO4 Formula Weight: 179.97 Melting Point: 136° MDL No.: MFCD02093046

Formula: C₈H₉BO₄
Melting Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD02093046
CAS Number: 99769-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR