5-Amino-2-methylpyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Amino-2-picoline, 6-Methylpyridin-3-amine, 3­Amino­6­methylpyridine, 6-Methyl-3-pyridinamine, 3­Amino­6­picoline

H27431-03 H27431-MD
AAH27431-03EA 198 USD
AAH27431-03 AAH27431-MD
5-Amino-2-methylpyridine 97%
5-Amino-2-methylpyridine

Purity: 97% CAS No.: 3430-14-6 Molecular Formula: C6H8N2 Formula Weight: 108.14 Odor: Characteristic Melting Point: 95-99° UN No.: 2811 MDL No.: MFCD00833389

Formula: C₆H₈N₂
MW: 108.14 g/mol
Melting Pt: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00833389
CAS Number: 3430-14-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR