Tricyclohexylphosphonium tetrafluoroborate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27428-03
H27428-06
H27428-14
AAH27428-03EA
30.4
USD
AAH27428-03
AAH27428-06
AAH27428-14
Tricyclohexylphosphonium tetrafluoroborate 99%
Tricyclohexylphosphonium tetrafluoroborate
Purity: 99%
CAS No.: 58656-04-5
Molecular Formula: C18H34P·BF4
Formula Weight: 368.25
Melting Point: 166-170°
MDL No.: MFCD03840580
Formula:
C₁₈H₃₄BF₄P Melting Pt: 166…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840580 CAS Number: 58656-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...