2,6-Dichloro-3-pyridineboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Dichloropyridin-3-ylboronic acid, 2,6-Dichloropyridine-3-boronic acid

H27406-MD H27406-03
AAH27406-MDEA 101 USD
AAH27406-MD AAH27406-03
2,6-Dichloro-3-pyridineboronic acid 95%
2,6-Dichloro-3-pyridineboronic acid

Purity: 95% CAS No.: 148493-34-9 Molecular Formula: C5H4BCl2NO2 Formula Weight: 191.81 MDL No.: MFCD03094989

Formula: C₅H₄BCl₂NO₂
MW: 191.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094989
CAS Number: 148493-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR