1-Butyl-4-methylpyridiniumhexafluorophosphate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Butyl-4-methylpyridinium Hexafluorophosphate
H27355-06
H27355-18
AAH27355-06EA
77.5
USD
AAH27355-06
AAH27355-18
1-Butyl-4-methylpyridiniumhexafluorophosphate 99%
1-Butyl-4-methylpyridiniumhexafluorophosphate
Purity: 99%
CAS No.: 401788-99-6
Molecular Formula: C10H16F6NP
Formula Weight: 295.22
Melting Point: 45°
MDL No.: MFCD03095438
Formula:
C₁₀H₁₆F₆NP MW: 295.21 g/mol Melting Pt: 45 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095438 CAS Number: 401788-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...