Lithium bis(trifluoromethylsulphonyl)imide 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lithium bis(trifluoromethanesulphonimide), Lithium bis((trifluoromethyl)sulphonyl)amide, Lithium Bis(Trifluoromethanesulphonyl)Imide, Bis(trifluoromethane)sulphonimide lithium salt
H27307-09
H27307-18
AAH27307-09EA
117.31
USD
AAH27307-09
AAH27307-18
Lithium bis(trifluoromethylsulphonyl)imide 98+%
Lithium bis(trifluoromethylsulphonyl)imide
Purity: 98+%
CAS No.: 90076-65-6
Molecular Formula: C2F6LiNO4S2
Formula Weight: 287.08
Melting Point: 234-238°
UN No.: 2923
MDL No.: MFCD00210017
Formula:
C₂F₆LiNO₄S₂ MW: 287.09 g/mol Melting Pt: 234…238 °C Storage Temperature: Ambient |
MDL Number:
MFCD00210017 CAS Number: 90076-65-6 EINECS: 415-300-0 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...