2,6-Diisopropyl-N,N-dimethylaniline 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27275-03
H27275-06
AAH27275-03EA
36.2
USD
AAH27275-03
AAH27275-06
2,6-Diisopropyl-N,N-dimethylaniline 96%
2,6-Diisopropyl-N,N-dimethylaniline
Purity: 96%
CAS No.: 2909-77-5
Molecular Formula: C14H23N
Formula Weight: 205.34
Flash Point:
Formula:
C₁₄H₂₃N Boiling Pt: 86 °C/2.5 mm Density: 0.873 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00144978 CAS Number: 2909-77-5 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...