4-Acetamido-2-methylbenzoic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27174-06
H27174-14
AAH27174-06EA
59.3
USD
AAH27174-06
AAH27174-14
4-Acetamido-2-methylbenzoic acid 96%
4-Acetamido-2-methylbenzoic acid
Purity: 96%
CAS No.: 103204-69-9
Molecular Formula: C10H11NO3
Formula Weight: 193.21
Melting Point: ca 248° dec.
MDL No.: MFCD02258874
Formula:
C₁₀H₁₁NO₃ MW: 193.21 g/mol Melting Pt: ∼ 248 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02258874 CAS Number: 103204-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...