2,6-Dimethoxyaniline 97%

Supplier: Thermo Scientific Chemicals
Danger

H27139-03 H27139-06
AAH27139-03EA 98.8 USD
AAH27139-03 AAH27139-06
2,6-Dimethoxyaniline 97%
2,6-Dimethoxyaniline

Purity: 97% CAS No.: 2734-70-5 Molecular Formula: C8H11NO2 Formula Weight: 153.18 Odor: Characteristic Melting Point: 76-78° UN No.: 2811 MDL No.: MFCD00053934

Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00053934
CAS Number: 2734-70-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR