1-Butyl-4-methylpyridiniumtetrafluoroborate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Butyl-4-methylpyridin-1-ium tetrafluoroborate
H27100-18
H27100-06
AAH27100-18EA
277
USD
AAH27100-18
AAH27100-06
1-Butyl-4-methylpyridiniumtetrafluoroborate 99%
1-Butyl-4-methylpyridiniumtetrafluoroborate
Purity: 99%
CAS No.: 343952-33-0
Molecular Formula: C10H16BF4N
Formula Weight: 237.06
Refractive Index: 1.4
Formula:
C₁₀H₁₆BF₄N MW: 237.05 g/mol Density: 1,2 Storage Temperature: Ambient |
MDL Number:
MFCD03095460 CAS Number: 343952-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...