4-tert-Butyl-1-(bromomethyl)benzene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(Bromomethyl)-4-(tert-butyl)benzene, 4-(tert-Butyl)benzylbromide

H26991-06 H26991-14
AAH26991-06EA 36.2 USD
AAH26991-06 AAH26991-14
4-tert-Butyl-1-(bromomethyl)benzene 97%
4-tert-Butyl-1-(bromomethyl)benzene

Purity: 97% CAS No.: 18880-00-7 Molecular Formula: C11H15Br Formula Weight: 227.14 Flash Point: 110°(230°F) Refractive Index: 1.5465 Melting Point: 8-12° Boiling Point: 93-94°/1.5mm Beilstein Reference: 471674 UN No.: 3265 MDL No.: MFCD00000180

Formula: C₁₁H₁₅Br
MW: 227.14 g/mol
Boiling Pt: 93…94 °C/1.5 mm
Melting Pt: 8…12 °C
Density: 1.236
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000180
CAS Number: 18880-00-7
EINECS: 242-643-1
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR