1-Butyl-3-methylpyridiniumtetrafluoroborate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Butyl-3-methylpyridin-1-ium tetrafluoroborate
H26983-06
H26983-18
AAH26983-06EA
96.9
USD
AAH26983-06
AAH26983-18
1-Butyl-3-methylpyridiniumtetrafluoroborate 99%
1-Butyl-3-methylpyridiniumtetrafluoroborate
Purity: 99%
CAS No.: 597581-48-1
Molecular Formula: C10H16BF4N
Formula Weight: 237.06
MDL No.: MFCD08458499
Formula:
C₁₀H₁₆BF₄N Density: 1,204 Storage Temperature: Ambient |
MDL Number:
MFCD08458499 CAS Number: 597581-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...