3-Iodopyridine-2-carbonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Iodopicolinonitrile, 2-Cyano-3-iodopyridine

H26963-03 H26963-MD
AAH26963-03EA 253 USD
AAH26963-03 AAH26963-MD
3-Iodopyridine-2-carbonitrile 95%
3-Iodopyridine-2-carbonitrile

Purity: 95% CAS No.: 827616-52-4 Molecular Formula: C6H3IN2 Formula Weight: 230.01 Melting Point: 98-100° UN No.: 3439 MDL No.: MFCD09037451

Formula: C₆H₃IN₂
MW: 230.01 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD09037451
CAS Number: 827616-52-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR