1-Methyl-3-phenylpiperazine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: MEFPIP

H26924-03 H26924-06 H26924-14
AAH26924-03EA 27.6 USD
AAH26924-03 AAH26924-06 AAH26924-14
1-Methyl-3-phenylpiperazine 97%
1-Methyl-3-phenylpiperazine

Purity: 97% CAS No.: 5271-27-2 Molecular Formula: C11H16N2 Formula Weight: 176.26 Melting Point: 56-60° Boiling Point: 85°/0.5mm UN No.: 2811 MDL No.: MFCD03411603

Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
Boiling Pt: 85 °C/0.5 mm
Melting Pt: 56…60 °C
Storage Temperature: Ambient
MDL Number: MFCD03411603
CAS Number: 5271-27-2
EINECS: 432-360-3
UN: 2923
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR