2-Chloro-5-ethylpyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

H26856-14 H26856-06
AAH26856-14EA 235 USD
AAH26856-14 AAH26856-03 AAH26856-06
2-Chloro-5-ethylpyrimidine 98%
2-Chloro-5-ethylpyrimidine

Purity: 98% CAS No.: 111196-81-7 Molecular Formula: C6H7ClN2 Formula Weight: 142.59 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Refractive Index: 1.521 Boiling Point: 223° MDL No.: MFCD00799503

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Boiling Pt: 223 °C
Density: 1.174
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00799503
CAS Number: 111196-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR