4-Phenylbenzonitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Biphenylcarbonitrile, 4-Cyanobiphenyl, p-Phenylbenzonitrile, [1,1'-Biphenyl]-4-carbonitrile
H26791-09
H26791-18
AAH26791-09EA
133
USD
AAH26791-09
AAH26791-18
4-Phenylbenzonitrile 95%
4-Phenylbenzonitrile
Purity: 95%
CAS No.: 2920-38-9
Molecular Formula: C13H9N
Formula Weight: 179.22
Flash Point:
Formula:
C₁₃H₉N Melting Pt: 85…87 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001821 CAS Number: 2920-38-9 EINECS: 220-860-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...