1-(4-Aminophenyl)-4-methylpiperazine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H26684-03
H26684-06
AAH26684-03EA
161
USD
AAH26684-03
AAH26684-06
1-(4-Aminophenyl)-4-methylpiperazine 97%
1-(4-Aminophenyl)-4-methylpiperazine
Purity: 97%
CAS No.: 16153-81-4
Molecular Formula: C11H17N3
Formula Weight: 191.27
Melting Point: 89-94°
UN No.: 3263
MDL No.: MFCD00172703
Formula:
C₁₁H₁₇N₃ Melting Pt: 89…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00172703 CAS Number: 16153-81-4 UN: 3263 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...