2,6-Difluoro-3-methoxyaniline 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Amino-2,4-difluoroanisole

H26631-03 H26631-06
AAH26631-03EA 202 USD
AAH26631-03 AAH26631-06
2,6-Difluoro-3-methoxyaniline 97%
2,6-Difluoro-3-methoxyaniline

Purity: 97% CAS No.: 144851-62-7 Molecular Formula: C7H7F2NO Formula Weight: 159.14 Refractive Index: 1.5235

Formula: C₇H₇F₂NO
Storage Temperature: Ambient
MDL Number: MFCD04115898
CAS Number: 144851-62-7
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR