4-Fluoro-2-methoxybenzonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26627-03
H26627-09
AAH26627-03EA
66.1
USD
AAH26627-03
AAH26627-09
4-Fluoro-2-methoxybenzonitrile 97%
4-Fluoro-2-methoxybenzonitrile
Purity: 97%
CAS No.: 191014-55-8
Molecular Formula: C8H6FNO
Formula Weight: 151.14
Melting Point: 75-77°
UN No.: 3439
MDL No.: MFCD04116335
Formula:
C₈H₆FNO Melting Pt: 75…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD04116335 CAS Number: 191014-55-8 UN: 3439 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...