3,5-Difluoro-4-methoxybenzyl bromide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H26526-03
H26526-06
AAH26526-03EA
210
USD
AAH26526-03
AAH26526-06
3,5-Difluoro-4-methoxybenzyl bromide 97%
3,5-Difluoro-4-methoxybenzyl bromide
Purity: 97%
CAS No.: 706786-42-7
Molecular Formula: C8H7BrF2O
Formula Weight: 237.04
Melting Point: 39-42°
UN No.: 3261
MDL No.: MFCD04115916
Formula:
C₈H₇BrF₂O Melting Pt: 39…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115916 CAS Number: 706786-42-7 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...