3-Formyl-6-methoxypyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methoxypyridine-3-carbaldehyde, 6-Methoxynicotinaldehyde, 6-Methoxypyridine-3-carboxaldehyde, 2-Methoxypyridine-5-carboxaldehyde, 6-Methoxy-3-pyridinecarboxaldehyde

H26515-06 H26515-03
AAH26515-06EA 239 USD
AAH26515-06 AAH26515-03
3-Formyl-6-methoxypyridine 98%
3-Formyl-6-methoxypyridine

Purity: 98% CAS No.: 65873-72-5 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Flash Point: 107°(225°F) Melting Point: 49-52° MDL No.: MFCD02683446

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 49…52 °C
Flash Pt: 107 °C(225 °F)
Storage Temperature: Ambient
MDL Number: MFCD02683446
CAS Number: 65873-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR