AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulfonylfluoride hydrochloride) 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
H26473-03
H26473-MD
AAH26473-03EA
750
USD
AAH26473-03
AAH26473-MD
AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulfonylfluoride hydrochloride) 97%
AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulphonylfluoride hydrochloride)
A covalent binding serine protease inhibitor
Purity: 97%
CAS No.: 30827-99-7
Molecular Formula: C8H10FNO2S.HCl
Formula Weight: 239.69
Storage Temperature: -30°C to -10°C
Melting Point: ca 184° dec.
Beilstein Reference: 7604627
UN No.: 3261
MDL No.: MFCD00132962
Formula:
C₈H₁₁ClFNO₂S MW: 239.69 g/mol Melting Pt: ∼ 184 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00132962 CAS Number: 30827-99-7 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...