4-Octylphenol 99%

Supplier: Thermo Scientific Chemicals
Danger

H26396-03 H26396-06
AAH26396-03EA 45.4 USD
AAH26396-03 AAH26396-06
4-Octylphenol 99%
4-Octylphenol

Purity: 99% CAS No.: 1806-26-4 Molecular Formula: C14H22O Formula Weight: 206.32 Flash Point: 110°(230°F) Melting Point: 43-45° Boiling Point: 150°/4mm UN No.: 2430 MDL No.: MFCD00036134

Formula: C₁₄H₂₂O
MW: 206.33 g/mol
Boiling Pt: 150 °C/4 mm
Melting Pt: 43…45 °C
Density: 0.961 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036134
CAS Number: 1806-26-4
EINECS: 217-302-5
UN: 2430
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR