α-Bromo-4-chloro-3-fluorotoluene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Bromomethyl)-1-chloro-2-fluorobenzene, 4-Chloro-3-fluorobenzyl bromide

H26391-03 H26391-06
AAH26391-03EA 114 USD
AAH26391-03 AAH26391-06
α-Bromo-4-chloro-3-fluorotoluene 97%
α-Bromo-4-chloro-3-fluorotoluene

Purity: 97% CAS No.: 206362-80-3 Molecular Formula: C7H5BrClF Formula Weight: 223.48 Melting Point: 30-33° UN No.: 3261 MDL No.: MFCD04115859

Formula: C₇H₅BrClF
MW: 223.47 g/mol
Melting Pt: 30…33 °C
Storage Temperature: Ambient
MDL Number: MFCD04115859
CAS Number: 206362-80-3
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR