3-(Trifluoromethyl)thiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H26311-03 H26311-MD
AAH26311-03EA 64.8 USD
AAH26311-03 AAH26311-06 AAH26311-MD
3-(Trifluoromethyl)thiobenzamide 97%
3-(Trifluoromethyl)thiobenzamide

Purity: 97% CAS No.: 53515-17-6 Molecular Formula: C8H6F3NS Formula Weight: 205.20 UN No.: 2811 MDL No.: MFCD01567258

Formula: C₈H₆F₃NS
MW: 151.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01567258
CAS Number: 53515-17-6
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR