4-Bromobenzyl mercaptan 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Bromotoluene-α-thiol

H26299-03 H26299-06 H26299-MD
AAH26299-03EA 118.69 USD
AAH26299-03 AAH26299-06 AAH26299-MD
4-Bromobenzyl mercaptan 98%
4-Bromobenzyl mercaptan

Purity: 98% CAS No.: 19552-10-4 Molecular Formula: C7H7BrS Formula Weight: 203.10 Flash Point: 110°(230°F) Refractive Index: 1.6170 Melting Point: 26-30° UN No.: 2811 MDL No.: MFCD03701566

Formula: C₇H₇BrS
MW: 203.1 g/mol
Melting Pt: 26…30 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03701566
CAS Number: 19552-10-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR