6-Chloro-m-toluidine 98%

Supplier: Thermo Scientific Chemicals
Warning

H26209-06 H26209-14
AAH26209-06EA 106 USD
AAH26209-06 AAH26209-14
6-Chloro-m-toluidine 98%
6-Chloro-m-toluidine

Purity: 98% CAS No.: 95-81-8 Molecular Formula: C7H8ClN Formula Weight: 141.60 Flash Point: 107°(225°F) Melting Point: 31-33° Boiling Point: 228-230° UN No.: 2239 MDL No.: MFCD00007674

Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 228…230 °C
Melting Pt: 31…33 °C
Flash Pt: 107 °C (225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007674
CAS Number: 95-81-8
EINECS: 202-454-7
UN: 2239
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR