6-Fluoro-m-toluonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

H26166-03 H26166-06
AAH26166-03EA 17.4 USD
AAH26166-03 AAH26166-06
6-Fluoro-m-toluonitrile 99%
6-Fluoro-m-toluonitrile

Purity: 99% CAS No.: 64113-84-4 Molecular Formula: C8H6FN Formula Weight: 135.14 Flash Point: 94°(201°F) Melting Point: 49-52° UN No.: 3439 MDL No.: MFCD00134542

Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 49…52 °C
Flash Pt: 94 °C (201 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134542
CAS Number: 64113-84-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR