2-Fluoro-6-methoxybenzyl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Bromomethyl)-1-fluoro-3-methoxybenzene

H26136-03 H26136-MD
AAH26136-03EA 118 USD
AAH26136-03 AAH26136-MD
2-Fluoro-6-methoxybenzyl bromide 98%
2-Fluoro-6-methoxybenzyl bromide

Purity: 98% CAS No.: 500912-17-4 Molecular Formula: C8H8BrFO Formula Weight: 219.05 Storage Temperature: 2°C to 8°C Refractive Index: 1.5570 Melting Point: 18-21° UN No.: 3265 MDL No.: MFCD08688579

Formula: C₈H₈BrFO
MW: 219.05 g/mol
Melting Pt: 18…21 °C
Storage Temperature: Ambient
MDL Number: MFCD08688579
CAS Number: 500912-17-4
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR