3-Fluoro-4-methoxybenzyl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Bromomethyl)-2-fluoro-1-methoxybenzene

H26116-03 H26116-MD
AAH26116-03EA 133 USD
AAH26116-03 AAH26116-MD
3-Fluoro-4-methoxybenzyl bromide 98%
3-Fluoro-4-methoxybenzyl bromide

Purity: 98% CAS No.: 331-61-3 Molecular Formula: C8H8BrFO Formula Weight: 219.05 Odor: Pungent Melting Point: 44-46° UN No.: 3261 MDL No.: MFCD00671768

Formula: C₈H₈BrFO
MW: 219.05 g/mol
Melting Pt: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00671768
CAS Number: 331-61-3
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR