4-Amino-3-fluorophenol 96%

Supplier: Thermo Scientific Chemicals
Danger

H26084-03 H26084-MD
AAH26084-03EA 180 USD
AAH26084-03 AAH26084-MD
4-Amino-3-fluorophenol 96%
4-Amino-3-fluorophenol

Purity: 96% CAS No.: 399-95-1 Molecular Formula: C6H6FNO Formula Weight: 127.12 Melting Point: 137-140° UN No.: 3077 MDL No.: MFCD00077452

Formula: C₆H₆FNO
Melting Pt: 137…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00077452
CAS Number: 399-95-1
EINECS: 402-230-0
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR