3-Pyrroline tech. 85% (remainder pyrrolidine), Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,5-Dihydro-1H-pyrrole
H26072-03
H26072-06
H26072-14
AAH26072-03EA
50.12
USD
AAH26072-03
AAH26072-06
AAH26072-14
3-Pyrroline tech. 85% (remainder pyrrolidine), Technical Grade
3-Pyrroline
Purity: 85%
CAS No.: 109-96-6
Molecular Formula: C4H7N
Formula Weight: 69.11
Storage Temperature: 2°C to 8°C
Flash Point: -18°(0°F)
Refractive Index: 1.4690
Odor: Unpleasant
Boiling Point: 90-91°
Beilstein Reference: 103173
Merck Reference: 14,8017
UN No.: 2733
MDL No.: MFCD00005213
Formula:
C₄H₇N MW: 69.11 g/mol Boiling Pt: 90 - 91 °C (1013 hPa) Melting Pt: -71 °C Density: 0.9097 g/cm³ (20 °C) Flash Pt: –18 °C (0 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005213 CAS Number: 109-96-6 EINECS: 203-723-1 UN: 2733 ADR: 3,III Merck Index: 12,08201 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...